missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli fabF Protein
A DNA sequence encoding the Escherichia coli (strain K12) fabF, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | NP_415613.1 |
---|---|
Gene ID (Entrez) | 946665 |
Name | fabF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088210
|
enQuireBio™
QP7326-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008220
|
enQuireBio™
QP7326-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098210
|
enQuireBio™
QP7326-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018220
|
enQuireBio™
QP7326-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028220
|
enQuireBio™
QP7326-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038220
|
enQuireBio™
QP7326-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_415613.1 | |
fabF Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
946665 | |
Research Use Only | |
fabF | |
Recombinant Protein | |
E. coli | |
SKRRVVVTGL GMLSPVGNTV ESTWKALLAG QSGISLIDHF DTSAYATKFA GLVKDFNCED IISRKEQRKM DAFIQYGIVA GVQAMQDSGL EITEENATRI GAAIGSGIGG LGLIEENHTS LMNGGPRKIS PFFVPSTIVN MVAGHLTIMY GLRGPSISIA TACTSGVHNI GHAARIIAYG DADVMVAGGA EKASTPLGVG GFGAARALST RNDNPQAASR PWDKERDGFV LGDGAGMLVL EEYEHAKKRG AKIYAELVGF GMSSDAYHMT SPPENGAGAA LAMANALRDA GIEASQIGYV NAHGTSTPAG DKAEAQAVKT IFGEAASRVL VSSTKSMTGH LLGAAGAVES IYSILALRDQ AVPPTINLDN PDEGCDLDFV PHEARQVSGM EYTLCNSFGF GGTNGSLIFK KI | |
Greater than 90% as determined by SDS-PAGE. |