missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Exopolyphosphatase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Exopolyphosphatase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7336-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088290

  • 3211.16 € / 1mg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_416997.1
Exopolyphosphatase Protein
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
946970
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ppx
Recombinant Protein
E. coli
PIHDKSPRPQ EFAAVDLGSN SFHMVIARVV DGAMQIIGRL KQRVHLADGL GPDNMLSEEA MTRGLNCLSL FAERLQGFSP ASVCIVGTHT LRQALNATDF LKRAEKVIPY PIEIISGNEE ARLIFMGVEH TQPEKGRKLV IDIGGGSTEL VIGENFEPIL VESRRMGCVS FAQLYFPGGV INKENFQRAR MAAAQKLETL TWQFRIQGWN VAMGASGTIK AAHEVLMEMG EKDGIITPER LEKLVKEVLR HRNFASLSLP GLSEERKTVF VPGLAILCGV FDALAIRELR LSDGALREGV LYEMEGRFRH QDVRSRTASS LANQYHIDSE QARRVLDTTM QMYEQWREQQ PKLAHPQLEA LLRWAAMLHE VGLNINHSGL HRHSAYILQN SDLPGFNQEQ QLMMATLVRY HRKAIKLDDL PRFTLFKKKQ FLPLIQLLRL GVLLNNQRQA TTTPPTLTLI TDDSHWTLRF PHDWFSQNAL VLLDLEKEQE YWEGVAGWRL KIEEESTPEI AA
Videos
SDS
Documents
Certificates
Promotions

Promotions