missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli DNA mismatch repair protein MutL Protein
A DNA sequence encoding the Escherichia coli (strain K12) DNA mismatch repair protein MutL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7511-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_418591.1 | |
DNA mismatch repair protein MutL Protein | |
Research Use Only | |
mutL | |
Recombinant Protein | |
E. coli | |
MPIQVLPPQL ANQIAAGEVV ERPASVVKEL VENSLDAGAT RIDIDIERGG AKLIRIRDNG CGIKKDELAL ALARHATSKI ASLDDLEAII SLGFRGEALA SISSVSRLTL TSRTAEQQEA WQAYAEGRDM NVTVKPAAHP VGTTLEVLDL FYNTPARRKF LRTEKTEFNH IDEIIRRIAL ARFDVTINLS HNGKIVRQYR AVPEGGQKER RLGAICGTAF LEQALAIEWQ HGDLTLRGWV ADPNHTTPAL AEIQYCYVNG RMMRDRLINH AIRQACEDKL GADQQPAFVL YLEIDPHQVD VNVHPAKHEV RFHQSRLVHD FIYQGVLSVL QQQLETPLPL DDEPQPAPRS IPENRVAAGR NHFAEPAARE PVAPRYTPAP ASGSRPAAPW PNAQPGYQKQ QGEVYRQLLQ TPAPMQKLKA PEPQEPALAA NSQSFGRVLT IVHSDCALLE RDGNISLLSL PVAERWLRQA QLTPGEAPVC AQPLLIPLRL KVSAEEKSAL EKAQSALAEL GIDFQSDAQH VTIRAVPLPL RQQNLQILIP ELIGYLAKQS VFEPGNIAQW IARNLMSEHA QWSMAQAITL LADVERLCPQ LVKTPPGGLL QSVDLHPAIK ALKDE | |
Greater than 80% as determined by SDS-PAGE. |
948691 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |