missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase RecQ Protein
A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase RecQ, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | WP_000035581.1 |
---|---|
Gene ID (Entrez) | 948318 |
Name | ATP-dependent DNA helicase RecQ Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057151
|
enQuireBio™
QP8915-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077151
|
enQuireBio™
QP8915-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067151
|
enQuireBio™
QP8915-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087151
|
enQuireBio™
QP8915-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097151
|
enQuireBio™
QP8915-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007161
|
enQuireBio™
QP8915-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000035581.1 | |
ATP-dependent DNA helicase RecQ Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
948318 | |
Research Use Only | |
recQ | |
Recombinant Protein | |
E. coli | |
AQAEVLNLES GAKQVLQETF GYQQFRPGQE EIIDTVLSGR DCLVVMPTGG GKSLCYQIPA LLLNGLTVVV SPLISLMKDQ VDQLQANGVA AACLNSTQTR EQQLEVMTGC RTGQIRLLYI APERLMLDNF LEHLAHWNPV LLAVDEAHCI SQWGHDFRPE YAALGQLRQR FPTLPFMALT ATADDTTRQD IVRLLGLNDP LIQISSFDRP NIRYMLMEKF KPLDQLMRYV QEQRGKSGII YCNSRAKVED TAARLQSKGI SAAAYHAGLE NNVRADVQEK FQRDDLQIVV ATVAFGMGIN KPNVRFVVHF DIPRNIESYY QETGRAGRDG LPAEAMLFYD PADMAWLRRC LEEKPQGQLQ DIERHKLNAM GAFAEAQTCR RLVLLNYFGE GRQEPCGNCD ICLDPPKQYD GSTDAQIALS TIGRVNQRFG MGYVVEVIRG ANNQRIRDYG HDKLKVYGMG RDKSHEHWVS VIRQLIHLGL VTQNIAQHSA LQLTEAARPV LRGESSLQLA VPRIVALKPK AMQKSFGGNY DRKLFAKLRK LRKSIADESN VPPYVVFNDA TLIEMAEQMP ITASEMLSVN GVGMRKLERF GKPFMALIRA HVDGD | |
Greater than 90% as determined by SDS-PAGE. |