missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 8-oxo-dGTP diphosphatase Protein

A DNA sequence encoding the Escherichia coli (strain K12) 8-oxo-dGTP diphosphatase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7301-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098070

  • 3405.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_414641.1
8-oxo-dGTP diphosphatase Protein
Research Use Only
mutT
Recombinant Protein
E. coli
MKKLQIAVGI IRNENNEIFI TRRAADAHMA NKLEFPGGKI EMGETPEQAV VRELQEEVGI TPQHFSLFEK LEYEFPDRHI TLWFWLVERW EGEPWGKEGQ PGEWMSLVGL NADDFPPANE PVIAKLKRL
Greater than 90% as determined by SDS-PAGE.
944824
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions