missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Annexin A13 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) Annexin A13, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8980-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087361

  • 1821.82 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001003255.1
Annexin A13 Protein
100 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
403935
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ANXA13
Recombinant Protein
Mammal
GNRHAKAKSH HGFDVDHDAK KLNKACKGMG TDEAAIIEIL SSRTSDERQQ IKQKYKATYG KDLEEVFKSD LSGNFEKTAL ALLDRPSEYD ARQLQKAMKG LGTDEAVLIE ILCTRTNKEI MAIKEAYQRL FDRSLESDVK ADTSGNLKAI LVSLLQANRD EGDDVDKDLA GQDAKDLYDA GDGRWGTDEL AFNEVLAKRS HKQLRATFQA YQILIDKDIE EAIEAETSGD LQKAYLTLVR CARDQEGYFA DRLYKSMKGT GTDEETLIHI IVTRAEVDLQ GIKAKFQEKY QKSLSDMVRS DTSGDFQKLL VALLH
Videos
SDS
Documents
Certificates
Promotions

Promotions