missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type C1 Protein

A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type C1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7104-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097390

Quantity Price
1 3116.48 € / 1mg
3 2910.17 € / 1mg
Estimated Shipment
09-06-2021
Add to basket


Description

Description

Specifications

Specifications

Botulinum neurotoxin type C1 Protein
1 mg
Not determined.
Recombinant Protein
E. coli
PITINNFNYS DPVDNKNILY LDTHLNTLAN EPEKAFRITG NIWVIPDRFS RNSNPNLNKP PRVTSPKSGY YDPNYLSTDS DKDPFLKEII KLFKRINSRE IGEELIYRLS TDIPFPGNNN TPINTFDFDV DFNSVDVKTR QGNNWVKTGS INPSVIITGP RENIIDPETS TFKLTNNTFA AQEGFGALSI ISISPRFMLT YSNATNDVGE GRFSKSEFCM DPILILMHEL NHAMHNLYGI AIPNDQTISS VTSNIFYSQY NVKLEYAEIY AFGGPTIDLI PKSARKYFEE KALDYYRSIA KRLNSITTAN PSSFNKYIGE YKQKLIRKYR FVVESSGEVT VNRNKFVELY NELTQIFTEF NYAKIYNVQN RKIYLSNVYT PVTANILDDN VYDIQNGFNI PKSNLNVLFM GQNLSRNPAL RKVNPEN
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Partial (see sequence information for more details).
C. botulinum
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents