missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine NucleoProtein
A DNA sequence encoding the Bos taurus (Bovine) Nucleoprotein, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Name | NucleoProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | N |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037660
|
enQuireBio™
QP7198-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057660
|
enQuireBio™
QP7198-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047660
|
enQuireBio™
QP7198-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067660
|
enQuireBio™
QP7198-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077660
|
enQuireBio™
QP7198-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087660
|
enQuireBio™
QP7198-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NucleoProtein | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
N | |
Recombinant Protein | |
E. coli | |
MALSKVKLND TFNKDQLLST SKYTIQRSTG DNIDIPNYDV QKHLNKLCGM LLITEDANHK FTGLIGMLYA MSRLGREDTL KILKDAGYQV RANGVDVITH RQDVNGKEMK FEVLTLVSLT SEVQGNIEIE SRKSYKKMLK EMGEVAPEYR HDFPDCGMIV LCVAALVITK LAAGDRSGLT AVIRRANNVL RNEMKRYKGL IPKDIANSFY EVFEKYPHYI DVFVHFGIAQ SSTRGGSRVE GIFAGLFMNA YGAGQVMLRW GVLAKSVKNI MLGHASVQAE MEQVVEVYEY AQKLGGEAGF YHILNNPKAS LLSLTQFPNF SSVVLGNAAG LGIMGEYRGT PRNQDLYDAA KAYAEQLKEN GVINYSVLDL TTEELEAIKN QLNPKDNDVE L | |
Greater than 90% as determined by SDS-PAGE. |