missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Mannose-binding protein 1

A DNA sequence encoding the Bos taurus (Bovine) Mannose-binding protein C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9267-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097951

  • 1100.00 € / 100µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_776532.1
Mannose-binding protein 1
Research Use Only
MBL
Recombinant Protein
Yeast
NCENIRKTCP VIACGPPGIN GIPGKDGRDG AKGEKGEPGQ GLRGSQGPPG KMGPQGTPGI PGIPGPIGQK GDPGENMGDY IRLATSERAT LQSELNQIKN WLIFSLGKRV GKKAFFTNGK KMPFNEVKTL CAQFQGRVAT PMNAEENRAL KDLVTEEAFL GITDQETEGK FVDLTGKGVT YQNWNDGEPN NASPGEHCVT LLSDGTWNDI ACSASFLTVC EFS
Greater than 90% as determined by SDS-PAGE.
281297
100 μg
Not determined.
Partial (see sequence information for more details).
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions