missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Interferon alpha-G Protein

A DNA sequence encoding the Bos taurus (Bovine) Interferon alpha-G, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7218-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097760

  • 2650.00 € / 500µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001165511.1
Interferon alpha-G Protein
Research Use Only
IFNAG
Recombinant Protein
Yeast
CHLPHTHSLA NRRVLTLLRQ LRRVSPSSCL QDRNDFAFPQ EALGGSQLQK AQAISVLHEV TQHTFQFFSV EGSAVVWDES LLDKLRDALD QQLTDLQFCL RQEEGLRGAP LLKEDSSLAV RKYFHRLTLY LQEKRHSPCA WEVVRAEVMR AFSSSTNLQE RFRRKD
Greater than 90% as determined by SDS-PAGE.
100329206
500 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions