missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Adiponectin / Acrp30 / ADIPOQ Protein

A DNA sequence encoding the Bos taurus (Bovine) Adiponectin / Acrp30 / ADIPOQ, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7647-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069990

  • 3211.16 € / 1mg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_777167.1
Adiponectin / Acrp30 / ADIPOQ Protein
1 mg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
282865
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADIPOQ
Recombinant Protein
E. coli
EDNMEDPPLP KGACAGWMAG IPGHPGHNGT PGRDGRDGTP GEKGEKGDPG LVGPKGDTGE TGITGIEGPR GFPGTPGRKG EPGESAYVYR SAFSVGLERQ VTVPNVPIRF TKIFYNQQNH YDGTTGKFLC NIPGLYYFSY HITVYLKDVK VSLYKNDKAL LFTHDQFQDK NVDQASGSVL LYLEKGDQVW LQVYEGENHN GVYADNVNDS TFTGFLLYHN IVE
Videos
SDS
Documents
Certificates
Promotions

Promotions