missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant B. abortus vjbR Protein
A DNA sequence encoding the Brucella abortus (strain 2308) vjbR, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | WP_002966460.1 |
---|---|
Gene ID (Entrez) | 29595784 |
Name | vjbR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029980
|
enQuireBio™
QP7642-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049980
|
enQuireBio™
QP7642-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039980
|
enQuireBio™
QP7642-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059980
|
enQuireBio™
QP7642-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069980
|
enQuireBio™
QP7642-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079980
|
enQuireBio™
QP7642-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_002966460.1 | |
vjbR Protein | |
Not determined. | |
Full Length | |
B. abortus | |
His | |
Tris-based buffer, 50% glycerol |
29595784 | |
Research Use Only | |
vjbR | |
Recombinant Protein | |
E. coli | |
MSLDLVHFPN YKKTFFGSSF QSDTLALLTR IRDEIGCRYV THTYRGRVGD CTKVNSADLT VLMTLPATWV ARYSSKNYFA IDPVFQEDAP YYRNDTSAIA RDLKEDADIC PAVAELLHDA EKHGLGNLFI AVSARNPKGV AGCTVFTFEV EDEDRTQFLA RMRPRLLSLA GIIHGTVCGC KDANSVASLL TPREVDCLRW AANGKTDGEI AEILSIARWT VVTYLQNAKI KLNCSNRTSA VATALSLGII DMPEVQHLV | |
Greater than 90% as determined by SDS-PAGE. |