missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein

A DNA sequence encoding the Acetobacter pasteurianus (Acetobacter turbidans) Modification methylase ApaLI, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7481-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088950

Quantity Price
1 3211.16 € / 1mg
3 3021.38 € / 1mg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

Modification methylase ApaLI Protein
1 mg
Not determined.
Full Length
A. pasturia
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
apaLIM
Recombinant Protein
E. coli
MNKDEVVVSL FAGAGGFSSG FSQAGLKPLF GAEINADACQ TYQENVGSPC HQLDLSTVDP SHIEMLTGGK RPFVVIGGPP CQGFSTAGPR NFADPRNLLI FNYLNIVERL SPRWLIFENV EGLLTSGGGR DLARLVREFV DMGYSVRLQK VNLAAYGVPQ TRKRVLIIGN RLGIDFQFPE ELYSFDSGKA KKASGKPLAP SLAEAVAGLG PAASDKDALV PYASSEPVNA FDARMRAGNR VEVVTHHVRV EAAERMQVEL LKPGQTMKDL PPELWHESYR RRANRRVSDG TPTEKRGGAP SGIKRLHGNL QSLTITGPAA REFIHPTEHR PLTIRECARI QTFPDKYRWV GNNASVIQQI GNAVPPLAAE RLAKHLRDID GSFGADTRPA GAMSAKLLGF VLTEALGMSP ALKSTEALLA EMHQGGFVF
Videos
SDS
Documents
Certificates
Promotions

Promotions