Learn More
Abnova™ RAD51 Recombinant Protein
Human RAD51 full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00005888-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are highly similar to bacterial RecA and Saccharomyces cerevisiae Rad51, and are known to be involved in the homologous recombination and repair of DNA. This protein can interact with the ssDNA-binding protein RPA and RAD52, and it is thought to play roles in homologous pairing and strand transfer of DNA. This protein is also found to interact with BRCA1 and BRCA2, which may be important for the cellular response to DNA damage. BRCA2 is shown to regulate both the intracellular localization and DNA-binding ability of this protein. Loss of these controls following BRCA2 inactivation may be a key event leading to genomic instability and tumorigenesis. Multiple transcript variants encoding different isoforms have been found for this gene
- Theoretical MW (kDa): 52.36
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MAMQMQLEANADTSVEEESFGPQPISRLEQCGINANDVKKLEEAGFHTVEAVAYAPKKELINIKGISEAKADKILAVAERYGLSGSDVLDNVAYARAFNTDHQTQLLYQASAMMVESRYALLIVDSATALYRTDYSGRGELSARQMHLARFLRMLLRLADEFGVAVVITNQVVAQVDGAAMFAADPKKPIGGNIIAHASTTRLYLRKGRGETRICKIYDSPCLPEAEAMFAINADGVGDAKD
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH01459 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
52.36 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MAMQMQLEANADTSVEEESFGPQPISRLEQCGINANDVKKLEEAGFHTVEAVAYAPKKELINIKGISEAKADKILAVAERYGLSGSDVLDNVAYARAFNTDHQTQLLYQASAMMVESRYALLIVDSATALYRTDYSGRGELSARQMHLARFLRMLLRLADEFGVAVVITNQVVAQVDGAAMFAADPKKPIGGNIIAHASTTRLYLRKGRGETRICKIYDSPCLPEAEAMFAINADGVGDAKD | |
BRCC5/HRAD51/HsRad51/HsT16930/RAD51A/RECA | |
RAD51 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
5888 | |
RAD51 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RAD51 | |
Human | |
Recombinant | |
Solution |