missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Upright Freezers, 32 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Marke: Tenak™ TE24130
Weitere Details : Gewicht : 3.10000kg
Beschreibung
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
Spezifikation
5 x 4 | |
Upright Freezers | |
Stainless Steel |
562 x 140 x 180 mm | |
Boxes (20) | |
32 mm (Box) |