missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Classic Rack for Chest Freezers, 130 mm Box Height
Classic Rack is suitable for use with all chest freezer models.
Marke: Tenak™ TE25102
Weitere Details : Gewicht : 1.16000kg
Beschreibung
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
Spezifikation
140 x 140 x 404 mm | |
130 mm (Box) | |
Locking rod |
Chest Frseezers | |
Boxes (3) | |
Stainless Steel |