Learn More
Tenak™ Rack
Brand:  Tenak™ TE21394
Additional Details : Weight : 0.95000kg
Product Code. 11597224
  • 71.55 € / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
freezer, side-up, for 3 boxes 50mm high, stainless steel, Tenak
Specifications
Specifications
Stainless Steel
1 x 3
420 mm
56 mm
Both chest and upright freezers
420 x 56 x 140 mm
140 mm
Cryoboxes (3, 50 mm H)