Learn More
R&D Systems™ Human Pluripotent Stem Cell 3-Color Immunocytochemistry Kit
To verify pluripotency in human stem cells using single-step ICC
Brand: R&D Systems™ SC021
Includes: 10X NL557-conjugated Goat Anti-Human SOX2, 10X NL637-conjugated Goat Anti-Human Oct-3/4, 10X NL493-conjugated Goat Anti-Human Nanog
Description
Defines the starting stem cell population
Increases confidence in pluripotent status by combining 3 different fluorochrome-conjugated antibodies for simultaneous staining
Reduces variability within an experiment and between studies
Allows researchers t
Specifications
1 Kit | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |