Learn More
(R)-4-(1-Aminoethyl)phenol, 97%, Thermo Scientific Chemicals
Catalog Number 15449428
Click to view available options
Quantity:
250 mg
1 g
5 g
134855-89-3
C8H12BrNO
218.094
MFCD03844647
PZBBMKOZPQAHRA-RGMNGODLSA-N
s-4-1-amino-ethyl-phenol hydrobromid, s---1-4-hydroxyphenyl ethylamine hydrobromide
49758827
4-[(1S)-1-aminoethyl]phenol;hydrobromide
CC(C1=CC=C(C=C1)O)N.Br
134855-89-3
C8H12BrNO
218.094
MFCD03844647
PZBBMKOZPQAHRA-RGMNGODLSA-N
s-4-1-amino-ethyl-phenol hydrobromid, s---1-4-hydroxyphenyl ethylamine hydrobromide
49758827
4-[(1S)-1-aminoethyl]phenol;hydrobromide
CC(C1=CC=C(C=C1)O)N.Br
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
UN Number UN3259
Formula Weight 137.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-4-(1-Aminoethyl)phenol