missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%, Thermo Scientific Chemicals

Catalog Number 15418576
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1212136-00-9
C19H21NO7S
407.437
MFCD09864994
RTHSATBSXDKZBN-QGZVFWFLSA-N
methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
46172874
methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC
This item is not returnable. View return policy
1212136-00-9
C19H21NO7S
407.437
MFCD09864994
RTHSATBSXDKZBN-QGZVFWFLSA-N
methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
46172874
methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC

Thiazoles are used in peptide research. Mainly used in medicine field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazoles are used in peptide research. Mainly used in medicine field.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.
TRUSTED_SUSTAINABILITY
Melting Point 100°C to 107°C
Color White to Brown
Quantity 250 mg
Formula Weight 237.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'