missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(1-Naphthyl)ethylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7057235
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3886-70-2
C12H13N
171.243
MFCD00064114
RTCUCQWIICFPOD-SECBINFHSA-N
r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine
2724264
(1R)-1-naphthalen-1-ylethanamine
CC(C1=CC=CC2=CC=CC=C21)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3886-70-2
C12H13N
171.243
MFCD00064114
RTCUCQWIICFPOD-SECBINFHSA-N
r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine
2724264
(1R)-1-naphthalen-1-ylethanamine
CC(C1=CC=CC2=CC=CC=C21)N

It is used in chiral synthesis in organic reactions including the synthesis of β-amino acids and the enantioselective of ketones to nitroolefins. Also used as absolute configuration of primary amines determined using chiral (2-nitrophenyl)proline amides and H NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in chiral synthesis in organic reactions including the synthesis of β-amino acids and the enantioselective of ketones to nitroolefins. Also used as absolute configuration of primary amines determined using chiral (2-nitrophenyl)proline amides and H NMR.

Solubility
Soluble in water (<10/l).

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.060 g/mL
Boiling Point 152°C to 153°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.623
Quantity 5 g
UN Number UN2922
Beilstein 2208025
Sensitivity Air sensitive
Solubility Information Soluble in water (<10/l).
Formula Weight 171.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-(1-Naphthyl)ethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'