Learn More
Sartorius Quintix™ Semi-Micro Balance with Internal Adjustment
The Quintix™ Semi-Micro Balance with Internal Adjustment makes your routine lab work easy with our revolutionary user interface.
Brand:  Sartorius QUINTIX125D-1CEU
Additional Details : Weight : 7.80000kg
Product Code. 16387978
  • 7900.00 € / Each
In Stock
to see stock.
Description
Description
  • Redefining StandardQuintix™ makes laboratory work more enjoyable.
  • The intuitive touch-screen in combination with precise weighing technology make Quintix™ laboratory balances a great choice for achieving the best results.
  • You will benefit from improved workflows by easy-to-use application programs.
  • If you need to generate GLP-compliant documents, you will save considerable time and effort with the ''plug and work'' connectivity of Quintix™ laboratory balances.
  • Intuitive operation of the balance without having to read a manual thanks to the new user interface and color touch screen Integrated application programs and intuitive user guidance for greater efficiency in your daily lab routine
  • Highest accuracy and convenience ensured at all times by the isoCAL fully automatic internal adjustment feature
  • Easier record-keeping using the balance's directly integrated PC functions and Plug and Work connectivity to a Sartorius lab printer
  • What's included:Universal power adapter, in-use cover, application guide, dust cover for analytical balances
Specifications
Specifications
CFR
Touchscreen
≤4.0 sec., 1.5 sec.
218 mm
60 to 120 g
0.01 mg, 0.1 mg
Balance