Learn More
Thermo Scientific™ QSP Filter and Non-Filtered Pipette Tips
Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Description
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.
- Micro point design tip orifice
- High clarity resin
- Graduation marks
- Universal fit
- Small micron filter
- Modern and robust tip rack design
- Non-Sterile:
- RNase/DNase
- DNA
- Pyrogen
- E-Beam Sterilized:
- RNase/DNase
- DNA
- Pyrogen/Endotoxin
- Bioburden
- PCR inhibitors
- Endotoxin ATP