missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyronin B, 50% dye content, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 174.0°C to 176.0°C |
Quantity | 5 g |
Beilstein | 18,V,10,182 |
Merck Index | 15,8118 |
Formula Weight | 521.11 |
Chemical Name or Material | Pyronin B |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'