missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,3-dicarboxylic anhydride, 98%, Thermo Scientific Chemicals

Catalog Number p-7028009
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
699-98-9
C7H3NO3
149.105
MFCD00005915
MCQOWYALZVKMAR-UHFFFAOYSA-N
2,3-pyridinedicarboxylic anhydride, furo 3,4-b pyridine-5,7-dione, quinolinic anhydride, 2,3-pyridinedicarboxylicanhydride, pyridine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyridine-5,7-dione, furano 3,4-b pyridine-5,7-dione, pyridine-2,3-dicarboxylic acid anhydride, 2,3-pyridinedicarboxylic anhydride quinolinic anhydride
69688
furo[3,4-b]pyridine-5,7-dione
C1=CC2=C(C(=O)OC2=O)N=C1
This item is not returnable. View return policy
699-98-9
C7H3NO3
149.105
MFCD00005915
MCQOWYALZVKMAR-UHFFFAOYSA-N
2,3-pyridinedicarboxylic anhydride, furo 3,4-b pyridine-5,7-dione, quinolinic anhydride, 2,3-pyridinedicarboxylicanhydride, pyridine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyridine-5,7-dione, furano 3,4-b pyridine-5,7-dione, pyridine-2,3-dicarboxylic acid anhydride, 2,3-pyridinedicarboxylic anhydride quinolinic anhydride
69688
furo[3,4-b]pyridine-5,7-dione
C1=CC2=C(C(=O)OC2=O)N=C1

Pyridine-2,3-dicarboxylic anhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2,3-dicarboxylic anhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Optimal solubility in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY
Melting Point 134°C to 138°C
Quantity 5 g
Beilstein 125111
Sensitivity Moisture sensitive
Solubility Information Optimal solubility in water.
Formula Weight 149.11
Percent Purity 98%
Chemical Name or Material Pyridine-2,3-dicarboxylic anhydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'