missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyridine-7-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7033064
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
362661-83-4
C8H6N2O
146.149
MFCD06739029
SGNBVQKSJWCGOJ-UHFFFAOYSA-N
pyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde, 8-hydropyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde 9ci, pyrazolo 1,5-alpha pyridine-7-carboxaldehyde
10866474
pyrazolo[1,5-a]pyridine-7-carbaldehyde
C1=CC2=CC=NN2C(=C1)C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
362661-83-4
C8H6N2O
146.149
MFCD06739029
SGNBVQKSJWCGOJ-UHFFFAOYSA-N
pyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde, 8-hydropyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde 9ci, pyrazolo 1,5-alpha pyridine-7-carboxaldehyde
10866474
pyrazolo[1,5-a]pyridine-7-carbaldehyde
C1=CC2=CC=NN2C(=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 83°C to 86°C
Quantity 250 mg
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material Pyrazolo[1,5-a]pyridine-7-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'