Learn More
Purified Human IgG(no IgA, IgM), MP Biomedicals
Contains no IgA or IgM
Brand:  MP Biomedicals 0855836
96.96 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : Weight : 0.00100kg
Product Code. 11474225
  • 103.56 € / 1mL
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Specifications
Unconjugated
1 mL