missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PTK6 (BRK), His Tag Recombinant Protein
Human PTK6 (BRK), His Tag Recombinant Protein
1826.00 € - 7890.00 €
Specifications
Accession Number | NP_005966 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 55.4 kDa |
Gene Alias | PTK6 (Brk) |
KinaseFamily | SRC Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15418403
|
Invitrogen™
PR4375B |
100μg |
1826.00 €
100µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10164842
|
Invitrogen™
PV4086 |
1 mg |
7890.00 €
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Breast tumor kinase (Brk) is expressed in over 60% of breast carcinoma tissue samples and breast tumor cell lines.
Specifications
NP_005966 | |
55.4 kDa | |
SRC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 200 mM NaCl, 1 mM EDTA, 0.05% Triton X-100, 5 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
PTK6 (Brk) | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus |