Learn More
SI Analytics™ PTFE Tubing Set
SI Analytics™ PTFE Tubing Set is suitable for thermostat types CT 53 and CT 54.
Brand:  SI Analytics™ 1060767
Additional Details : Weight : 0.80000kg
Product Code. 10610963
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • PTFE tubing set pressure VZ 8521
  • 1.5mm inner diameter
  • Suitable to thermostat type: CT 53 und CT 54
Specifications
Specifications
Tubing Set
1.5mm Diameter
For Thermostat Type CT 53, CT 54, CT 72/2 and CT 72/4
Thermostat Type CT 53, CT 54, CT 72/2 and CT 72/4
PTFE