Learn More
Lenz Laborglasinstrumente™ Stirrers with PTFE Blade and Journal
Ideal for routine stirring and mixing applications. Lenz Laborglasinstrumente™ Stirrers with PTFE Blade and Journal feature a shaft manufactured from durable DURAN™ tubing.
Brand: Lenz Laborglasinstrumente™ B3972557
Additional Details : Weight : 0.06300kg
Description
- Precision calibrated glass tube
- Ground and polished bearing surface
- With snap fastener for replacement of PTFE blades