Learn More
VITLAB™ PTFE Oval Magnetic Stirring Bar
Brand:  VITLAB™ 311797
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10000691
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Permanent magnet AlNiCo V core
For preparation, separation and concentration
PTFE
Oval
20 mm
3/Pk.
50 mm