Learn More
Bohlender™ PTFE Egg Shape Magnetic Stirring Bar
Brand:  Bohlender™ C358-08
Additional Details : Weight : 0.01800kg
 View more versions of this product
Product Code. 10053261
Quantity Price
1 13.68 € / Each
10 11.44 € / Each
In stock, ready to ship: 1
Add to basket
You may only order up to the available stock. For more details call customer services on 0800-20 88 40.
Description
Description
Specifications
Specifications
For stirring in round bottom flasks, Shape mimics that of the flasks and assures complete mixing.
15mm
PTFE (Polytetrafluoroethylene)
Egg
Alnico 5 magnetic core
35mm
1/Pk.