Learn More
Bohlender™ PTFE Cylindrical Magnetic Stirring Bar
Brand:  Bohlender™ C350-08
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10013401
Quantity Price
1 1.53 € / Each
10 1.43 € / Each
In stock, ready to ship: 17
Add to basket
You may only order up to the available stock. For more details call customer services on 0800-20 88 40.
Specifications
FDA Certified
Alnico 5 magnetic core
10mm
1/Pk.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
3mm
PTFE (Polytetrafluoroethylene)
Cylindrical