missing translation for 'onlineSavingsMsg'
Learn More

Propargyltrimethylsilane, 80-90%, stabilized, Thermo Scientific Chemicals

Catalog Number p-2692098
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13361-64-3
C6H12Si
112.25
MFCD00042922
ULYLMHUHFUQKOE-UHFFFAOYSA-N
propargyltrimethylsilane, silane, trimethyl-2-propynyl, trimethyl 2-propynyl silane, trimethyl prop-2-ynyl silane, trimethyl propargyl silane, silane, trimethyl-2-propyn-1-yl, trimethyl prop-2-yn-1-yl silane, trimethyl-2-propynylsilane, propargyl trimethylsilane, 2-propynyltrimethylsilane
83378
trimethyl(prop-2-ynyl)silane
C[Si](C)(C)CC#C
This item is not returnable. View return policy
13361-64-3
C6H12Si
112.25
MFCD00042922
ULYLMHUHFUQKOE-UHFFFAOYSA-N
propargyltrimethylsilane, silane, trimethyl-2-propynyl, trimethyl 2-propynyl silane, trimethyl prop-2-ynyl silane, trimethyl propargyl silane, silane, trimethyl-2-propyn-1-yl, trimethyl prop-2-yn-1-yl silane, trimethyl-2-propynylsilane, propargyl trimethylsilane, 2-propynyltrimethylsilane
83378
trimethyl(prop-2-ynyl)silane
C[Si](C)(C)CC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 0.7500g/mL
Boiling Point 91°C to 93°C
Flash Point -10°C
Infrared Spectrum Authentic
Assay Percent Range 80 to 90% (GC)
Packaging Glass bottle
Linear Formula HC∽CCH2Si(CH3)3
Quantity 1 g
Beilstein 04, IV, 3938
Specific Gravity 0.75
Solubility Information Solubility in water: insoluble
Stabilizer 0.1 wt% hydroquinone
Formula Weight 112.25
Percent Purity 80-90%
Physical Form Liquid
Chemical Name or Material Propargyltrimethylsilane, 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Propargyltrimethylsilane, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'