missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Procarbazine hydrochloride

Catalog Number 15527729
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
366-70-1
C12H19N3O·HCl
257.76
DERJYEZSLHIUKF-UHFFFAOYSA-N
procarbazine hydrochloride, procarbazine hcl, matulane, nathulane, natunalar, mih hydrochloride, pcb hydrochloride, procarbazine.hcl, ibenzmethyzine hydrochloride, natulan hydrochloride
9703
CHEBI:71428
4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride
CC(C)NC(=O)C1=CC=C(C=C1)CNNC.Cl
This item is not returnable. View return policy
366-70-1
C12H19N3O·HCl
257.76
DERJYEZSLHIUKF-UHFFFAOYSA-N
procarbazine hydrochloride, procarbazine hcl, matulane, nathulane, natunalar, mih hydrochloride, pcb hydrochloride, procarbazine.hcl, ibenzmethyzine hydrochloride, natulan hydrochloride
9703
CHEBI:71428
4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride
CC(C)NC(=O)C1=CC=C(C=C1)CNNC.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Loss on Drying 2% max.
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 257.76
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Procarbazine hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Procarbazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'