missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPM1B (Human) Recombinant Protein
Human PPM1B full-length ORF ( AAH12002, 1 a.a. - 192 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | AAH12002 |
---|---|
Gene ID (Entrez) | 5495 |
Name | protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139411
|
Abnova™
H00005495-P01.10UG |
10 μg |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149411
|
Abnova™
H00005495-P01.25UG |
25 μg |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MSIVLVCFSNAPKVSDEAVKKDSELDKHLESRVEEIMEKSGEEGMPDLAHVMRILSAENIPNLPPGGGLAGKRNVIEAVYSRLNPHRESDGASDEAEESGSQGKLVEALRQMRINHRGNYRQLLEEMLTSYRLAKVEGEESPAEPAATATSSNSDAGNPVTMQESHTESESGLAELDSSNEDAGTKMSGEKI
Specifications
AAH12002 | |
protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform | |
125% SDS-PAGE Stained with Coomassie Blue | |
MGC21657, PP2C-beta-X, PP2CB, PP2CBETA, PPC2BETAX | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5495 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPM1B | |
GST |