Learn More
Dostmann Electronic Power Pack Adaptor for P600-Series Modem
Robust, reliable power pack adapter for P600-series modem.
Brand: Dostmann Electronic 5600-0004
Additional Details : Weight : 0.20000kg
Description
- Power Pack for P600-series 230 VAC
- Includes adaptor for RS232-cable
- 9 V adapter for P500/P600
Order Info
VoltageRange
Specifications
Power Pack Adaptor | |
P600-Series Modem | |
1000 mm | |
1 Ea. |