missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Potential-Free Contact (24V/2A) W/Socket To Namur Ne 28 For Combined Error Message, For Uf260Ts
Brand: Memmert™ UF260TSH6
Additional Details : Weight : 0.01100kg
Includes: Socket 3-Pin
Specifications
Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108 | |
Potential-free Contact | |
24 V | |
2 A |