missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN110

Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).

Brand:  Memmert™ UN110H6

Additional Details : Weight : 0.01200kg

Product Code. 17599775

  • 277.00 € / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

  • Accessory
  • For combination error message
Specifications

Specifications

UN110 Oven
Socket to NAMUR NE 28
2 A
Potential-Free Contact
24 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions