missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, BAKER ANALYZED™ ACS, J.T. Baker™
399.00 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Specifications
Boiling Point | 1330°C (1013 hPa) |
Melting Point | 686°C |
pH | 6.0 to 9.2 (5% Solution at 25°C) |
Quantity | 500 g |
Assay Percent Range | min. 99.0% |
Loss on Drying | max. 0.2% (at 150°C) |
Density | 3.13 g/cm³ (20°C) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'