missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-cyanophenyltrifluoroborate, 96%, Thermo Scientific™

Catalog Number p-7035470
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
850623-36-8
C7H4BF3KN
209.02
MFCD04115736
JEQAKQWNUUSKHI-UHFFFAOYSA-N
potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanopheny trifluoroborate, potassium 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyltrifluoroborat, trifluoro 4-cyanophenyl potassioboron v, potassium 4-cyanophenyl-trifluoroboranuide, potassium ion 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoro borate 1-
23716872
potassium;(4-cyanophenyl)-trifluoroboranuide
[K+].F[B-](F)(F)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
850623-36-8
C7H4BF3KN
209.02
MFCD04115736
JEQAKQWNUUSKHI-UHFFFAOYSA-N
potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanopheny trifluoroborate, potassium 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyltrifluoroborat, trifluoro 4-cyanophenyl potassioboron v, potassium 4-cyanophenyl-trifluoroboranuide, potassium ion 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoro borate 1-
23716872
potassium;(4-cyanophenyl)-trifluoroboranuide
[K+].F[B-](F)(F)C1=CC=C(C=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 1 g
Formula Weight 209.02
Percent Purity 96%
Chemical Name or Material Potassium 4-cyanophenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'