missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-carboxyphenyltrifluoroborate, 95%, Thermo Scientific™

Catalog Number p-7034218
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
850313-91-6
C7H5BF3KO2
228.019
MFCD07784423
LKEMWADDENRLDB-UHFFFAOYSA-N
potassium 3-carboxyphenyltrifluoroborate, potassium 3-carboxyphenyl trifluoroborate, potassium 3-carboxyphenyl trifluoroboranuide, pubchem11349, amtb109, potassium 3-carboxyphenyltrifluoroborat, potassium 3-carboxyphenyl trifluoro borate 1-
23682740
potassium;(3-carboxyphenyl)-trifluoroboranuide
[B-](C1=CC(=CC=C1)C(=O)O)(F)(F)F.[K+]
This item is not returnable. View return policy
850313-91-6
C7H5BF3KO2
228.019
MFCD07784423
LKEMWADDENRLDB-UHFFFAOYSA-N
potassium 3-carboxyphenyltrifluoroborate, potassium 3-carboxyphenyl trifluoroborate, potassium 3-carboxyphenyl trifluoroboranuide, pubchem11349, amtb109, potassium 3-carboxyphenyltrifluoroborat, potassium 3-carboxyphenyl trifluoro borate 1-
23682740
potassium;(3-carboxyphenyl)-trifluoroboranuide
[B-](C1=CC(=CC=C1)C(=O)O)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 228.02
Percent Purity 95%
Chemical Name or Material Potassium 3-carboxyphenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'