Learn More
Alfa Aesar™ Potassium 3-carboxyphenyltrifluoroborate, 95%
Brand:  Alfa Aesar™ H51885.03
Additional Details : CAS : 850313-91-6 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15418387
  • 38.17 € / 1g
In stock, ready to ship: 1
Add to basket
Description
Description
Specifications
Chemical Identifiers
850313-91-6
228.019
LKEMWADDENRLDB-UHFFFAOYSA-N
23682740
[B-](C1=CC(=CC=C1)C(=O)O)(F)(F)F.[K+]
C7H5BF3KO2
MFCD07784423
potassium 3-carboxyphenyltrifluoroborate, potassium 3-carboxyphenyl trifluoroborate, potassium 3-carboxyphenyl trifluoroboranuide, pubchem11349, amtb109, potassium 3-carboxyphenyltrifluoroborat&, potassium 3-carboxyphenyl trifluoro borate 1-
potassium;(3-carboxyphenyl)-trifluoroboranuide
Specifications
Potassium 3-carboxyphenyltrifluoroborate
850313-91-6
MFCD07784423
LKEMWADDENRLDB-UHFFFAOYSA-N
potassium;(3-carboxyphenyl)-trifluoroboranuide
23682740
95%
1g
C7H5BF3KO2
potassium 3-carboxyphenyltrifluoroborate, potassium 3-carboxyphenyl trifluoroborate, potassium 3-carboxyphenyl trifluoroboranuide, pubchem11349, amtb109, potassium 3-carboxyphenyltrifluoroborat&, potassium 3-carboxyphenyl trifluoro borate 1-
[B-](C1=CC(=CC=C1)C(=O)O)(F)(F)F.[K+]
228.019
228.02