Learn More
Gilson™ Position Rack Tube Vessel Type, Liquid Handlers
These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments
Brand:  Gilson™ 150424
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16656115
  • 199.92 € / Each
Estimated Shipment: 07-01-2022
Add to basket
Description
Description
  • The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates
  • Tubes Vessel Type
Specifications
Specifications
44
Code 22
25 mL 18 x 150 mm Vial
Tubes