Learn More
Mettler Toledo™ Portamess™ Portable Conductivity Meters
Brand:  Mettler Toledo™ 99606123
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 10077841
  • 1205.00 € / Each
Estimated Shipment: 15-11-2021
Add to basket
Description
Description
Specifications
Specifications
Clock, datalogger, interface, software
Portamess Conductometer
-20°C to +120°C
Portamess 913 Cond
0.01 μS/cm to 1000 mS/cm