missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, M.W. 40,000, Thermo Scientific Chemicals

Catalog Number p-7042401
Click to view available options
Quantity:
100 g
500 g
1 kg
9003-39-8
(C6H9NO)n
111.14
MFCD01076626
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
*-CC(-*)N1CCCC1=O
9003-39-8
(C6H9NO)n
111.14
MFCD01076626
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
*-CC(-*)N1CCCC1=O

A cryoprotectant, also useful in plant cell culture research

A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Merck Index 14,7697
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 40 kd
Chemical Name or Material Polyvinylpyrrolidone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'