Learn More
Greiner Bio-One Round Bottom Polystyrene Test Tubes
Clear, round bottom test tubes for sample transfer.
Brand: Greiner Bio-One 160101
Additional Details : Weight : 11.44000kg
Description
Clear, round bottom test tubes for sample transfer.
Polystyrene tubes are ideally suited for optical measurements as a result of its high clarity.
- Nonsterile
- Available in different sizes
Service Plan
No
Specifications
Clear | |
100 mm | |
Sample transfer | |
16 mm |
12 mL | |
Polystyrene | |
Non-sterile | |
Test Tube |