Learn More
VITLAB™ Polystyrene Sterile Pipets
Designed for convenient and easy sterile pipetting. VITLAB™ Polystyrene Sterile Pipets are disposable, graduated pipets that are individually wrapped with an included filter. They are constructed of crystal clear, pyrogen-free polystyrene and are available in a variety of volumes.
Brand:  VITLAB™ 160210
Additional Details : Weight : 0.13500kg
 View more versions of this product
Product Code. 15211539
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
VITLAB™ pipettes are individually measured volumetrically during production, and are provided with a calibration mark. These graduated pipettes feature nominal volume at the bottom and complete run-out only for the nominal volume. They are disposable and ready to use for lab applications.
  • Crystal clear and with a printed scale for easy reading of partial volumes
  • Includes a cotton-wool filter
  • Pre-sterilized and individually packaged
  • Constructed of pyrogen-free polystyrene
    • Light weight and easy to use
    • High break resistance
    • Disposable and convenient
  • Manufactured to the glass pipette standard
    • Correspond to Class B in accordance with DIN EN ISO 835
    • Identified by bar-code
Specifications
Specifications
Polystyrene
0.01 mL
Crystal Clear
Pipet
Yes
25 Pack
2 mL
Single-use