Learn More
Witeg™ Polypropylene Screw Caps
Catalog Number p-8008791
Click to view available options
Color:
Neutral
Yellow
Use Witeg™ Polypropylene Screw Caps to keep contents inside.your laboratory bottles. Various colors help you categorize your items.
  • Autoclavable up to 140°C
  • GL 45 Thread
Closure Material Polypropylene
Closure Size GL 45
Product Type Screw Cap
Color Yellow
Material Polypropylene
No. per Pack 10Pack
Quantity 10Pack
Screw Color Yellow
Screw Material Polypropylene
Closure Color Yellow
Autoclavable Autoclavable