Learn More
Brady™ Polypropylene Mandatory Signs
13.25 € - 32.30 €
Specifications
Material | Polypropylene |
---|---|
Quantity | 1 Ea. |
Shape | Round |
Product Type | Mandatory Sign |
Color | White on Blue |
Product Code | Brand | Displayed Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Price | Quantity & Availability | |||||
10315724
|
Brady™
252300 |
Commandment Icon - Cradle Net Compulsory |
13.25 €
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
10745345
|
Brady™
252109 |
Commandment Icon - Respiratory Protection Required |
13.25 €
Each |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
10235034
|
Brady™
252128 |
Commandment Icon - Safety Helmet and Eye Protection |
32.30 €
Each |
Estimated Shipment: 02-05-2024
Log in to see stock. |
|||||
10000864
|
Brady™
252126 |
Commandment Icon - Safety Helmet and Ear Protection Compulsory |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Polypropylene | |
Round | |
White on Blue |
1 Ea. | |
Mandatory Sign |